Learn More
Tocris Bioscience™ Bosentan
High affinity dual ETA and ETB receptor antagonist;orally bioavailable
Brand: Tocris Bioscience™ 6232/50
3351.10 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Bosentan is a high affinity dual ETA and ETB receptor antagonist (Ki values are 4.7 and 95 nM for ETA and ETB, respectively). Inhibits ET-1 induced contraction of isolated rat aorta and sarafotoxin S6C -induced contraction of rat trachea in vitro. Orally bioavailable.Specifications
Antagonist | |
147536-97-8 | |
551.61g/mol | |
Non-selective Endothelin Receptor Antagonists |
Non-selective Endothelin Receptor | |
C27H29N5O6S | |
Store at 4°C | |
50 mg |
For Research Use Only