Learn More
Borradaile Needle
Brand:  S Murray SAMCO D210/01
Product Code. 12784406
  • 326.00 NOK / Pack of 10
Only null left
Estimated Shipment: 04-08-2025
Specifications
Borradaile Needle
Stainless Steel
30 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Borradaile Needle > Length: 30mm