Learn More
Lenz Laborglasinstrumente™ Borosilicate Tower Pycnometers
Tower Pycnometers constructed of borosilicate glass.
668.00 NOK - 907.00 NOK
Specifications
Quantity | 1 Ea. |
---|---|
Type | Tower Pycnometer |
Material | Borosilicate Glass |
Product Type | Tower Pycnometer |
Product Code | Brand | Neck Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Neck Size | Price | Quantity & Availability | ||||||
10010491
|
Lenz Laborglasinstrumente™
05660207 |
NS 60/27 |
907.00 NOK
Each |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||||
10758841
|
Lenz Laborglasinstrumente™
05660206 |
NS 45/27 |
668.00 NOK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
||||||
Description
- Borosilicate Glass
Specifications
1 Ea. | |
Tower Pycnometer | |
Borosilicate Glass | |
Tower Pycnometer |
Product Title
Lenz Laborglasinstrumente™ Borosilicate Tower Pycnometers