Learn More
Fisherbrand™ Borosilicate Glass Wide Neck Erlenmeyer Flasks With Graduation
Brand: Fisherbrand™ SPFS66650-3000
649.69 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Specifications
3000 mL | |
Borosilicate Glass | |
Erlenmeyer Flask | |
Wide Neck |
Promotions
-
Multiple Manufacturers
The Academic Best Buy Book 2025
Browse exclusive prices for our academic customersPromo Code: 25081Expires: 2025-12-31
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13