Learn More
Fisherbrand™ Borosilicate Glass Test Tube
Store test reagents and materials. Fisherbrand™ borosilicate glass test tubes come in a variety of sizes.
348.00 NOK - 466.00 NOK
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | |||||
15872275
|
Fisherbrand™
90106120 |
75 mm | 12 mm |
348.00 NOK
Pack of 100 |
Estimated Shipment: 30-07-2025
Log in to see stock. |
|||||
15822285
|
Fisherbrand™
90106141 |
150 mm | 24 mm | N/A | ||||||
Description
- Borosilicate glass test tubes, grade 3.3
- All ISO 4142
Specifications
ISO 4142 | |
Borosilicate Glass |
Product Title
Fisherbrand™ Borosilicate Glass Test Tube