Learn More
Borosilicate Glass T-shape Stopcock Adapter
Control flow rate with a T-shape adapter. The QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
Brand:  Quickfit MF11/3B/SC
 View more versions of this product
Product Code. 10349804
  • 738.00 NOK / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Description
  • With GP Rotaflo™ PTFE stopcock providing fine flow control and positive closure
  • Detachable plastic screw thread connectors allow rubber hose etc. , to be fitted easily and safely
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
T-shape
Stopcock Adapter
24/29
GP Rotaflo™ PTFE stopcock