Learn More
Kimble Chase™ Borosilicate Glass Shell Vials, No Cap
Short-style shell vials are ideal for storing dry products
Brand:  Kimble Chase™ 60931-6
966.49 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 11843193
  • 1105.00 NOK / Pack of 144
Price-Shattering Deals on Lab Glassware
Use promo code 24123 at checkout
View Available Promotions
Only null left
Estimated Shipment: 25-06-2025
Description
  • Plain top design
  • Lab pack quantities
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 mL
576
144
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kimble Chase™ Borosilicate Glass Shell Vials, No Cap > Capacity: 24mL