Learn More
Kimble Chase™ Borosilicate Glass Shell Vials, No Cap
Short-style shell vials are ideal for storing dry products
Brand: Kimble Chase™ 60931-14
377.15 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Plain top design
- Lab pack quantities
- Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Specifications
1 mL | |
1440 | |
144 |
KIM001 Kimble Chase 08 09 Catalog
File size: 57.1 MB
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
Product Content Correction
Product Title
Kimble Chase™ Borosilicate Glass Shell Vials, No Cap > Capacity: 1mL