Learn More
BRAND™ Borosilicate Glass Powder Funnel
Manufactured from borosilicate glass. BRAND™ Borosilicate Glass Powder Funnel includes ground and fire-polished upper rim. Short, wide stem is optimal for funneling powder and other ground materials.
Brand:  BRAND™ 146523
369.67 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 10623592
  • 470.00 NOK / Each
Price-Shattering Deals on Lab Glassware
Use promo code 24123 at checkout
View Available Promotions
Only null left
Estimated Shipment: 09-07-2025
Description
  • Manufactured from borosilicate glass.
  • Ground and fire-polished upper rim.
  • Short, wide stem optimal for funneling powder and other ground materials.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
120 mm
30 mm
Borosilicate Glass
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Borosilicate Glass Powder Funnel > Top Diameter: 120 mm