Learn More
Quickfit™ Borosilicate Glass Pear Shaped Claisen-Vigreux Flask
Incorporates short, Vigreux type fractionating column.
Brand:  Quickfit MD4/50
Product Code. 15507930
Description
Description
  • Column indents aid intimate contact between vapor and liquid
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pear
50 mL
Borosilicate Glass
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Pear Shaped Claisen-Vigreux Flask > Capacity: 50mL