Learn More
Kimble Chase™ Borosilicate Glass Gas Sampling Bulbs
Brand:  Kimble Chase™ 653150-0125
 View more versions of this product
Product Code. 11740409
  • 3670.00 NOK / Each
Specifications
Volume: 125 mL
Documents
One moment while we fetch your results.
Product Certifications
Promotions
Promotions
Product Content Correction
Product Title
Kimble Chase™ Borosilicate Glass Gas Sampling Bulbs > Volume: 125mL