Learn More
IKA™ Borosilicate Glass Coated Receiving Flask
Brand:  IKA™ 0003743200
 View more versions of this product
Product Code. 11895932
  • 1836.71 NOK / Each
Estimated Shipment: 29-08-2025
Specifications
Borosilicate Glass
Receiving Flask
1 Each
100 mL
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
IKA™ Borosilicate Glass Coated Receiving Flask > Capacity: 100mL