Learn More
Pyrex™ Borosilicate Glass Automatic Burette with Intermediate Stopcock
2705.00 NOK - 2930.00 NOK
Products 3
Product Code Brand Capacity (Metric) Graduations Price Quantity & Availability  
Product Code Brand Capacity (Metric) Graduations Price Quantity & Availability  
12832104
View Documents
Pyrex
3496/04AS
25 mL 0.05 mL
2815.00 NOK
Each
Only null left
Estimated Shipment: 06-10-2025
Add to basket
12842104
View Documents
Pyrex
3496/06AS
50 mL 0.1 mL
2930.00 NOK
Each
Only null left
Estimated Shipment: 06-10-2025
Add to basket
12822104
View Documents
Pyrex
3496/02AS
10 mL 0.02 mL
2705.00 NOK
Each
Only null left
Estimated Shipment: 06-10-2025
Add to basket
Description
Description
  • As 3396 range with intermediate stopcock
  • Intermediate stopcock facilitates easy drainage of liquid back into reservoir
  • 1.5mm bore PTFE key stopcock
Documents
Product Certifications
Product Content Correction
Product Title
Pyrex™ Borosilicate Glass Automatic Burette with Intermediate Stopcock