Learn More
BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper, Hexagonal Grip
Hollow with drip tip and hexagonal grip. BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper is constructed from Boro 3.3 to DIN 12252 standards.
Brand:  BRAND™ 144605
54.37 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 10141441
  • 64.80 NOK / Each
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Description
  • Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
  • Hollow
  • Hexagonal grip
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1/Pk.
NS 7/16
Ground Joint Stopper
NS 7/16
Borosilicate Glass
Conical
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    30
    Hours
    13
    Minutes
    2
    Seconds
    13
Product Content Correction
Product Title
BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper, Hexagonal Grip > NS 7/16