Learn More
Bohlender™ BOLA™ Tri-clamp GL Stopcocks, DN 08
Tri-clamp, GL 2-way stopcock with straight bore.
Brand:  Bohlender™ D917-02
Product Code. 17157334
  • 1208.40 NOK / Each
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 2-way stopcock made of PTFE with straight bore
  • One Tri-Clamp neck as per DIN 32 676 and one GL-threaded neck
  • Cylindrical stopcock made of PTFE for reliable tightness and grip made of PP for marking the flow direction
  • Universal chemical resistance
  • Product is only exposed to PTFE
Specifications
Specifications
DN 08
25 mm
1 in.
Thread GL 18
6 mm
PTFE
8 mm
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Product Certifications
Offerte speciali
Offerte speciali