Learn More
Bohlender™ BOLA™ Tri-clamp Gaskets
Double-sided tri-clamp gaskets with elastic PTFE sleeve and silicone inlay.
Brand:  Bohlender™ D930-26
 View more versions of this product
Product Code. 17117344
  • 232.00 NOK / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Silicone ring with double-sided, elastic washer made of PTFE
  • Suitable for Tri-clamp connections as per DIN 32 676
  • Universal chemical resistance
  • Product is only exposed to PTFE
Specifications
Specifications
Tubing with 20 mm I.D.
1.3 in.
PTFE, Silicone
DN 20
34 mm