Learn More
Bohlender™ BOLA™ Balls
Balls for use as splash guards or extending surfaces.
Brand: Bohlender™ H964-03
This item is not returnable.
View return policy
Description
- Made of solid PTFE, with smooth surface
- Different packing units
- For extension of surfaces such as distillation apparatus or as splash guard
Specifications
PTFE | |
3 mm | |
Smooth Surface | |
100 Pk. |
Product Content Correction
Product Title
Bohlender™ BOLA™ Balls > Balls; Diameter (Metric): 3 mm; Material: PTFE; Quantity: 100 Pk.; Description: Smooth Surface