missing translation for 'onlineSavingsMsg'
Learn More

BOC-L-alaninal, 98%

CAS: 79069-50-4 | C8H15NO3 | 173.21 g/mol

Brand:  Thermo Scientific Acros 438470010

2481.71 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Quantity 1 g

Product Code. 11905491

  • 2495.00 NOK / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

86.4°C to 87.6°C
White to Yellow
97.5% min. (GC)
Glass Bottle
-30.6
OEQRZPWMXXJEKU-LURJTMIESA-N
tert-butyl N-[(2S)-1-oxopropan-2-yl]carbamate
1 g
173.21
Powder
79069-50-4
Authentic
C8H15NO3
MFCD00143786
boc-l-alaninal, boc-ala-aldehyde, s-tert-butyl 1-oxopropan-2-yl carbamate, boc-l-alanine aldehyde, tert-butyl n-2s-1-oxopropan-2-yl carbamate, s-tert-butyl 1-oxopropan-2-ylcarbamate, chembl94727, s-1-methyl-2-oxo-ethyl-carbamic acid tert-butyl ester, carbamic acid, 1s-1-methyl-2-oxoethyl-, 1,1-dimethylethyl ester, 1-methyl-2-oxo-ethyl-carbamic acid tert-butyl ester
C[C@H](NC(=O)OC(C)(C)C)C=O
173.21
7015728
98%
BOC-L-Alaninal
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Boc-L-alaninal

SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number341288 Certificate TypeCertificate of Analysis Date04/12/2023 Catalog Number438470010
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BOC-L-alaninal, 98% > 1g; Glass bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.