missing translation for 'onlineSavingsMsg'
Learn More

BOC-D-Alanine, 99+%, Thermo Scientific Chemicals

546.88 NOK - 1876.65 NOK

Chemical Identifiers

CAS 7764-95-6
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00063123
InChI Key QVHJQCGUWFKTSE-RXMQYKEDSA-N
Synonym boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid
PubChem CID 637606
SMILES C[C@@H](NC(=O)OC(C)(C)C)C(O)=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10247020
View Documents Promotion Details
Thermo Scientific Acros
275720050
5 g Glass Bottle
546.88 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
10666174
View Documents Promotion Details
Thermo Scientific Acros
275720250
25 g Glass Bottle
1876.65 NOK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7764-95-6
189.21
QVHJQCGUWFKTSE-RXMQYKEDSA-N
637606
C8H15NO4
MFCD00063123
boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid
C[C@@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

81°C to 84°C
White
C8H15NO4
(CH3)3COC(O)nHC(CH3)COOH
04, 2507
QVHJQCGUWFKTSE-RXMQYKEDSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
189.21
189.21
Powder
7764-95-6
99+%
Authentic
MFCD00063123
boc-d-alanine, boc-d-ala-oh, n-boc-d-alanine, n-tert-butoxycarbonyl-d-alanine, r-2-tert-butoxycarbonyl amino propanoic acid, n-alpha-t-boc-d-alanine, boc-d-ala, n-t-butoxycarbonyl-d-alanine, 2r-2-tert-butoxy carbonyl amino propanoic acid
C[C@@H](NC(=O)OC(C)(C)C)C(O)=O
+25° to +27° (20°C, 589nm) (c=2, CH3COOH)
637606
≥99.0%
BOC-D-Alanine
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only