missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP5 Mouse anti-Human, Porcine, Clone: 1G6C10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66370-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. These proteins are synthesized as prepropeptides, cleaved, and then processed into dimeric proteins. This protein may act as an important signaling molecule within the trabecular meshwork and optic nerve head, and may play a potential role in glaucoma pathogenesis. This gene is differentially regulated during the formation of various tumors.Specifications
BMP5 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P22003 | |
BMP5 | |
BMP5 Fusion Protein Ag3952 | |
150 μL | |
Primary | |
Human, Porcine | |
Antibody | |
IgG1 |
Western Blot | |
1G6C10 | |
Unconjugated | |
BMP5 | |
BMP 5, BMP5, bone morphogenetic protein 5 | |
Mouse | |
Protein G | |
RUO | |
100151964, 653 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction