Learn More
Brady™ BMP21-Plus Label Printer Kit
Brand: Brady™ 134462
Additional Details : Weight : 2.22800kg Transport : UN number : 3481 Chem class : 9
Specifications
BMP21-PLUS Portable Label Printer | |
Thermal Transfer | |
235 mm | |
19 mm | |
BMP21-PLUS Label Printer - Electrical Printer Kit - EU | |
Croatian, Czech, Danish, Dutch, English, Estonian, Finnish, French, German, Hungarian, Italian, Norwegian (Bokmål), Polish, Portuguese, Romanian, Slovak, Slovenian, Spanish, Swedish, Turkish | |
10 mm/sec. | |
Li-ion Battery and EU Power Cord | |
0.75 kg | |
1 Ea. | |
6 AA Batteries, Rechargeable Battery, AC Adaptor |
LCD | |
125 mm | |
50 mm | |
Monochrome | |
Li-ion Battery and a EU Power Cord | |
Saves 12 files | |
203 dpi | |
5°C to 40°C | |
Bin and shelf identification, Lean identification, Patch panel marking, Rating Plates, Terminal block marking, Wire and cable marking, General and industrial marking, Voice and data communication | |
2 Years |