Learn More
Sampling Systems™ Blue SteriWare™ Scrapers
BIOPHARMA
The risk of cross contamination is eliminated by using Blue SteriWare™ Scrapers. Cleanroom manufactured and available sterile, these funnels are ideal for scraping and cleaning in the food, cosmetic, and pharmaceutical industry. Blue color allows you to see the scraper if left in product.
Brand:  Sampling Systems™ 8785B-75
 View more versions of this product
Product Code. 17584686
  • 2110.00 NOK / Pack of 100
Only null left
Estimated Shipment: 07-08-2025
Description
  • This range of blue disposable Scrapers are available sterile and non-sterile.
  • They are made from high quality polymers.
  • These scrapers are ideal in the manufacture of creams, ointments and pastes.
  • The bright blue color means they can easily seen if they are left in the product by mistake.
  • SteriWare™ Scrapers are available sterile and non-sterile.
  • Moulded and packed in a cleanroom.
  • Individually bagged.
  • Made form FDA and EU conforming materials.
  • BSE/TSE free.
  • Full Batch Traceability.
  • Available as gamma irradiated sterile (SAL 10-6).
Specifications
Specifications
Non-sterile
75 x 75 mm
Polypropylene
Individually Bagged
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number000844 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number8785B-75
Lot Number00844 Certificate TypeCertificate of Compliance Date14/10/2024 Catalog Number8785B-75
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ Blue SteriWare™ Scrapers > Blue Scraper; Dimensions (L x W): 75 x 75 mm; Color: Blue; Sterility: No; Material: Polypropylene