Learn More
Buerkle™ Blue Detectable Forceps
Blue detachable forceps feature a special additive in the material that makes them visible to metal detectors or X rays.
Brand: Buerkle™ 5378-1060
This item is not returnable.
View return policy
Description
- Suitable for precise lab work and are ideal for grasping, lifting and moving small objects
- The fluted handles ensure optimum and secure handling as well as precise work
- The light plastic forceps for single use allow for fatigue-free work
- The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined
- The lab tweezers are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses
- Individually packaged
- For single use
- Clean room manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
- Pointed/angled tip: The disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends
- Pointed tip: The anatomical forceps have a straight tip, fluted on the inside. This makes it possible to grip sensitive materials reliably
- Broad tip: The disposable forceps have ends with a broad tip. The broad tip is smooth and blunt. It prevents the material from being damaged when it is gripped
Specifications
Pointed, Angled | |
5.12 in. | |
130 mm | |
Sterile |
100 | |
Polystyrene | |
Forceps |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Brochure Disposable samplers for foodstuffs
File size: 535.2 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
245032 | Certificate of Analysis | 12/02/2025 | 5378-1060 |
243096 | Certificate of Compliance | 21/08/2024 | 5378-1060 |
234085 | Certificate of Compliance | 27/09/2023 | 5378-1060 |
221721 | Certificate of Compliance | 23/05/2022 | 5378-1060 |
224489 | Declaration of Conformity | 07/12/2022 | 5378-1060 |
Product Content Correction
Product Title
Buerkle™ Blue Detectable Forceps > Tip Style: Pointed, Angled