Learn More
Karl Hecht™ Blood Smear Slides
An excellent accessory for the microscopic examination of blood samples.
Brand:  Karl Hecht™ 40619010
Product Code. 15844802
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • According to Undritz
  • CE marking
  • Clear glass
  • Both sides polished
  • Beveled edges and corners
  • Thickness from 1.6 to 1.7mm
  • Dimensions approx. 40 x 20mm
Specifications
Specifications
Undritz blood smear slide
Microscopic blood examination