Learn More
Thermo Scientific™ Oxoid™ Blood agar base powder Oxoid
Brand: Thermo Scientific™ Oxoid™ CM0055T
11901.21 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Straw powder | |
General purpose | |
5 kg |
Powder | |
7.1 to 7.5 |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Thermo Scientific™ Oxoid™ Blood agar base powder Oxoid >