Learn More
BRAND™ Blaubrand™ One-Mark Bulb Pipettes
Unique class AS volumetric pipettes made of Soda-lime glass with 1 marking and blue graduation with a volume of 20ml, a tolerance of 0.03 ±ml, and an overall length of 510 ± 10mm.
Brand:  BRAND™ 29704
586.11 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11323854
  • 605.00 NOK / Pack of 6
Elevate Your Beverage Testing with Precision Solutions
Use promo code 24135 at checkout
View Available Promotions
Only null left
Estimated Shipment: 16-07-2025
Description
  • Volumetric pipettes, class AS, Soda-lime glass, 1 marking, blue graduation
  • Blaubrand™ DIN EN ISO 648
  • DE-M marking with batch number
  • Graduations and inscriptions in highly contrasting blue enamel
  • Calibrated to deliver (EX)
  • All Blaubrand™ volumetric pipettes are supplied with batch number and one batch certificate per manufacturer's outer pack (up to 2ml: 12 cylinders, above 2ml: 6 cylinders per pack)
  • Also available with individual certificate or DAkkS (former DKD) calibration certificate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6/Pk.
2.5 mL
One-mark Bulb Pipet
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Blaubrand™ One-Mark Bulb Pipettes > Capacity: 2.5mL; Packaging: 6/pk