Learn More
Brand™ BLAUBRAND™ Class A Volumetric Flask with Flanged Rim, with Certificate
Manufactured from Borosilicate Glass and complies with DIN EN ISO 1042 standards.BRAND™ Blaubrand™ Class A Volumetric Flask with Flanged Rim prevents slippage when suspended by a clamp. Batch number and individual certificate included. Delivered with individual conformity certificate.
Brand: Brand™ 937047
This item is not returnable.
View return policy
Description
- Manufactured from Borosilicate Glass.
- Beaded rim prevents slippage when suspended by a clamp.
- Complies with DIN EN ISO 1042 standards.
- Calibrated to contain (TC, In.)
- Batch number and individual certificate included.
- Delivered with individual conformity certificate.
Specifications
Calibrated To Contain | |
Clear | |
Borosilicate Glass | |
2 Piece | |
A |
DIN EN ISO 1042, DakkS calibration certificate available upon request. | |
25 mL | |
±0.04 mL | |
±0.04 mL |