Learn More
mb2 Chair on Glides - Black Fabric Upholstered - Asynchronous Mechanism
Click to view available options
Color:
Black
Unit Size:
Each
Description
- Chair in fabric black
- Seat and back in blue fabric upholstered
- Asynchronous mecanism, adjustable seat and backrest
- Pneumatic cylinder Mid
- 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
- Glides
Specifications
Specifications
Product Type | Contact Seat Chair |
Color | Black |
Adjustable (Height) | Yes |
Adjustable | Height |
Back Style | Ergonomic |
Height (Metric) | 660 mm |
Holds | 5 Pads |
Seating Height | 470 to 690 mm |
Weight (Metric) | 10.22 kg |
With Armrest | No |
Show More |
Product Content Correction
Product Title
mb2 Chair on Glides - Black Fabric Upholstered - Asynchronous Mechanism >