Learn More
BRAND™ BISTABIL™ Bottles For Automatic Burettes
Narrow-mouth bottle with tube bushing
Brand: BRAND™ 126965
350.95 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
2000 mL | |
Clear Soda-Lime Glass | |
Gas Washing Bottle | |
Automatic Burettes |
Promotions
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
Product Content Correction
Product Title
BRAND™ BISTABIL™ Bottles For Automatic Burettes > Capacity (Metric): 2000 mL