Learn More
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals Product Code.: 10629004
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals
Product Code. 10629004
25 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Product Code. 10629004
Brand: Thermo Scientific Acros 387600250
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 82113-65-3
Molecular Formula C2HF6NO4S2
Molecular Weight (g/mol) 281.16
MDL Number MFCD00214154
InChI Key ZXMGHDIOOHOAAE-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-n-trifluoromethyl sulfonyl methanesulfonamide, trifluoromethanesulfonimide, unii-c7r849vm9l, bis trifluoromethanesulfonyl imide, bistrifluoromethanesulfonimide, 1,1,1-trifluoro-n-trifluoromethanesulfonylmethanesulfonamide, 1,1,1-trifluoro-n-trifluoromethylsulfonyl methanesulfonamide, bis trifluoromethane sulfonimide, n,n-bis trifluoromethanesulfonyl imide, n,n-bis trifluoromethylsulphonyl amineShow MoreShow Less
PubChem CID 157857
IUPAC Name 1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
SMILES C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F
Specifications
Melting Point 52°C to 56°C
Color Brown to Colorless
Boiling Point 90°C to 91°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: reacts
Formula Weight 281.16
Percent Purity 99%
Physical Form Crystals
Chemical Name or Material Bis(trifluoromethanesulfonyl)imide
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466008 Certificate TypeCertificate of Analysis Date12/09/2024 Catalog Number387600010,387600250,387600050
Lot NumberA0459475 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number387600010,387600250,387600050
Lot NumberA0441294 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog Number387600010,387600250,387600050
Lot NumberA0455540 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number387600010,387600250,387600050
Lot NumberA0451026 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number387600010,387600250,387600050
compliance-icons
Product Identifier
  • Bis(trifluoromethanesulfonyl)imide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Product Title
Bis(trifluoromethanesulfonyl)imide, 99%, Thermo Scientific Chemicals >