Learn More
BioWhittaker™ DMEM high glucose with UltraGlutamine
Sustain growth for most types of cells. BioWhittaker™ DMEM high glucose with UltraGlutamine is suitable for, human, monkey, hamster, rat, mouse, chicken and fish cells.
Brand: Lonza BE12-604F/U1
This item is not returnable.
View return policy
Description
- DMEM is a modification of Basal Medium Eagle (BME)
- Contains four-times the concentrations of the amino acids and vitamins.
- Manufactured in accordance with cGMP regulations
- The formulations used for standard liquid media products are those recommended by the Tissue Culture Association
Specifications
cGMP | |
Cell Culture Media | |
500 mL |
4.5g/L | |
Mammalian Cell Culture |