Learn More
Lonza™ BioWhittaker™ General-Purpose Serum-Free Media
Chemically defined media optimized for specific cell types
7293.00 NOK
Specifications
Content And Storage 2° to 8°C
View More Specs
Products 1
Product Code Brand Product Type Quantity Price Quantity & Availability  
Product Code Brand Product Type Quantity Price Quantity & Availability  
11660121
View Documents
Lonza™
77232
Serum Free Medium (SFM) 2 x 500 mL N/A N/A N/A
Description
Description
PC-1™
  • Provides complete media system, including PC-1 Supplement for cultivation of primary cells and anchorage-dependent cell lines
  • Contains complete HEPES buffering system
UltraCULTURE™
  • Designed for growth of adherent and nonadherent cells
Specifications
Specifications
2° to 8°C
Documents
Product Certifications
Product Content Correction
Product Title
Lonza™ BioWhittaker™ General-Purpose Serum-Free Media