Learn More
Kinematica Biotrana™ High-Turbulence Mixer for PT3100 D
High-turbulence mixing head with the least possible shear forces and energy input.
Brand:  Kinematica 11030314
Product Code. 15761876
  • 34830.00 NOK / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Description
  • Selection and applications:
    • Quick removal and suspension of solid particles; also suitable for high viscosity applications
Specifications
Specifications
34 m/s
36 mm
High-turbulence Mixer
1 Ea.
500 mL to 10 L
250 mm
PT-DA 36/BEC-F25