missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bioreactor Probe Assembly for use with Single-Use Bioreactors

Bioreactor Probe Assembly for HyPerforma™ and DynaDrive™ Single-use Bioreactors.

21060.00 NOK - 29810.00 NOK

Specifications

Autoclavable Autoclavable
Material Silicone
For Use With HyPerforma™ Single-use Bioreactors, DynaDrive™ Single-use Bioreactors
Color Clear
Product Type Probe Assembly
View More Specs

Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
30040280
View Documents
Thermo Scientific™
SH30720.02
Kleenpak Connector
21060.00 NOK
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
30040274
View Documents
Thermo Scientific™
SH30720.03
Aseptiquick G Connector
29810.00 NOK
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The Thermo Scientific™ Bioreactor Probe Assembly (nonsterile) is used to package the pH and DO probes for sterilization and to aseptically connect them to the BPCs for Thermo Scientific™ Single-Use Bioreactors.
Specifications

Specifications

Autoclavable
HyPerforma™ Single-use Bioreactors, DynaDrive™ Single-use Bioreactors
Probe Assembly
Silicone
Clear
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Bioreactor Probe Assembly for use with Single-Use Bioreactors

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.