Learn More
Sartorius™ Biohit™ Tip Ejector Collars
Allows safe and easy pipette tip removal without touching the tips by hand. Sartorius™ Biohit™ Tip Ejector Collars are designed for use with a variety of mechanical and electronic single- or multi-channel pipette models/sizes.
162.05 NOK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11512804
|
Sartorius™
721054 |
10μL, 20μL, 50μL, 100μL, 200μL Tips of Proline Mechanical Pipette | N/A | ||||||
Description
- Removes pipette tips without touching the tips by hand
- Activates a spring loaded tip cone
- Tips are ejected with a simple thumb press on the ejector button
- Compatible with Biohit™ tips, including Optifit tips and SafetySpace™ Filter tips
- Available in several sizes for a variety of mechanical and electronic units
- Made of durable thermoplastic
Specifications
Tip Ejector Collar |