Learn More
Ansell BioClean™ Durableu Disposable Overshoes BDBO
Click to view available options
Size:
Standard
X-Large
Description
- Resists tears, rips, and abrasions
- Low levels of particle shedding
- Resistant to a wide range of liquid chemicals
- High tensile strength
- Slip-resistant
- Non-woven spunbonded polypropylene (38 gsm)/Cast polyethylene laminate (64 gsm)
Specifications
Specifications
Size | Standard |
Material | Polypropylene |
Latex Free | No |
Color | Blue |
Disposable | Single-use |
Sterility | Non-sterile |
Certifications/Compliance | Manufactured in a facility holding ISO 9001:2015 |
Packaging Quantity | 50 Pieces per Sealed Inner PE Bag, 1 Inner Bag per Sealed Outer PE Bag, 8 Outer Bags per Lined Carton |
Packaging Type | 400/Cs. |
Product Type | Controlled Environments Shoe Covers |
Five (5) years from date of manufacture. Store in a dry, cool place (≤40°C) away from direct sunlight and fluorescent light.
Product Content Correction
Product Title
Ansell BioClean™ Durableu Disposable Overshoes BDBO >