Learn More
Ansell BioClean™ Durableu Disposable Overboots BDBL
Click to view available options
Size: 420 mm (16.5 in.)
Description
- Resists tears, rips and abrasions
- Low levels of particle shedding
- Resistant to a wide range of liquid chemicals
- High tensile strength
- Slip-resistant
- 102gsm non-woven, spunbonded Polypropylene (38gsm)/Cast Polyethylene (64gsm) laminate
Specifications
Specifications
Size | Universal |
Latex Free | No |
Color | Blue |
Disposable | Single-use |
Sterility | Non-sterile |
Certifications/Compliance | Manufactured in a facility holding ISO 9001:2015 |
Packaging Quantity | 50 pieces per sealed inner PE bag; one inner bag per sealed outer PE bag; eight outer bags per lined carton (400 pieces) |
Resistance | Chemical-resistant |
Product Type | Controlled Environments Boot Covers |
Videos
-
Nitritex Bioclean-D
Five (5) years from date of manufacture. Store in a dry, cool place (≤40°C) away from direct sunlight and fluorescent light.
Product Content Correction
Product Title
Ansell BioClean™ Durableu Disposable Overboots BDBL >