Learn More
Ansell BioClean-D™ Sterile Hood
Long length cleanroom hood features an extra-long yoke for maximum coverage.
Brand: Ansell S-BDHD-L
2612.87 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Alert:
N/A
Description
- Extended length: More complete protective coverage
- Low-linting lightweight material: Fewer contamination risks
- ESD properties: Reduced risk of electrostatic interference or damage
Specifications
Sterile Hood | |
Polypropylene | |
Pull-over | |
No | |
Bound | |
Universal | |
Low-linting, Lightweight Material | |
Electrostatic Discharge |
Universal | |
White | |
Single-use | |
S-BDHD | |
Universal | |
Sterile | |
Laboratory, Crime Scene Investigation (CSI), Forensics, Cleanroom, Medical Device Manufacturing, Biotechnology, Maintaining Equipment and Instruments, Pharmaceutical | |
Class 10 (ISO 4) |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
24-0055 | Certificate of Irradiation | 16/05/2024 | S-BDHD-L |
24-0055 | Certificate of Compliance | 16/05/2024 | S-BDHD-L |
20-0857 | Certificate of Compliance | 15/12/2022 | S-BDHD-L |
20-0857 | Certificate of Sterility | 15/12/2022 | S-BDHD-L |
18-3743 | Certificate of Sterility | 15/12/2022 | S-BDHD-L |