Learn More
Thermo Scientific™ BioBasic™ 8 PEEK Bio-Inert HPLC Columns, 5μm Particle Size
Thermo Scientific™ BioBasic 8 PEEK Bio-Inert HPLC Columns, 5μm Particle Size, are optimized for the separation of a wide range of peptides.
Brand: Thermo Scientific™ 72205-252168
Description
Thermo Scientific™ BioBasic 8 PEEK Bio-Inert HPLC Columns, 5μm Particle Size, are optimized for the separation of a wide range of peptides. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules. Optimized for the separation of medium hydrophobicity peptides, nominally less than 20,000 daltons.
- 300Ă… pore size for improved biomolecule analysis
- An excellent starting column for protein and peptide analysis
- Outstanding reproducibility, efficiency and column lifetime
- Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available in bio-inert column hardware. Please call Customer Service for more information.

Specifications
PEEK Column | |
LC/MS | |
5 ÎĽm | |
300 Ă… | |
L7 | |
BioBasic | |
5% | |
Yes |
2.1 mm | |
250 mm | |
2 to 9 | |
100 m²/g | |
C8 | |
Silica | |
Reversed Phase | |
Reversed Phase |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.