missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bilirubin, 98+%

Bilirubin, CAS # 635-65-4, is a tetrapyrroliccompound found in bile that is a product of heme catabolism. | CAS: 635-65-4 | C33H36N4O6 | 584.67 g/mol

Chemical Identifiers

CAS 635-65-4
Molecular Formula C33H36N4O6
Molecular Weight (g/mol) 584.67
MDL Number MFCD00005499
InChI Key BPYKTIZUTYGOLE-IFADSCNNSA-N
Synonym bilirubin, hematoidin, hemetoidin, bilirubin ix-alpha, principal bile pigment, unii-rfm9x3lj49, bilirubin ixalpha, 21h-biline-8,12-dipropanoic acid, 2,17-diethenyl-1,10,19,22,23,24-hexahydro-3,7,13,18-tetramethyl-1,19-dioxo, rfm9x3lj49, biline-8,12-dipropionic acid, 1,10,19,22,23,24-hexahydro-2,7,13,17-tetramethyl-1,19-dioxo-3,18-divinyl
PubChem CID 5280352
ChEBI CHEBI:16990
IUPAC Name 3-[2-[[3-(2-carboxyethyl)-5-[(Z)-(3-ethenyl-4-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-2-yl]methyl]-5-[(Z)-(4-ethenyl-3-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-3-yl]propanoic acid
SMILES CC1=C(C=C)\C(NC1=O)=C\C1=C(C)C(CCC(O)=O)=C(CC2=C(CCC(O)=O)C(C)=C(N2)\C=C2/NC(=O)C(C=C)=C2C)N1
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Bilirubin is a degradation product of heme following the activity of heme oxidase. Bilirubin is produced in the reticuloendothelial system and conjugated in the liver. It appears as an orange-yellow pigment that is secreted in the bile
  • High levels of unconjugated bilirubin can result in jaundice, a condition of the yellowing of skin and eyes. This can be toxic and may be a sign of severe liver disease
  • Bilirubin has been seen to have functions including cell signaling, metabolism modulation, and immune regulation
  • It has protective antioxidant activity at moderate levels

Application

  • Bilirubin is widely used in studies of liver function and disease, such as hepatitis or cirrhosis
  • As an antioxidant, bilirubin has been studied in protection against H2O2-mediated cell death
  • It can be used as a scavenger of reactive oxygen species (ROS) or as an inhibitor of NADPH oxidase
Specifications
Show More
Videos
SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Bilirubin, 98+%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.