missing translation for 'onlineSavingsMsg'
Learn More
Bicarbonate, 1M buffer soln., pH 8.0, Thermo Scientific™ Product Code.: 15415719

Bicarbonate, 1M buffer soln., pH 8.0, Thermo Scientific™

Product Code. 15415719
250 mL
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy

Product Code. 15415719

Brand: Thermo Scientific Alfa Aesar J63491.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Concentration 1 M
pH 8.0
Physical Form Liquid
Quantity 250 mL
Chemical Name or Material Bicarbonate buffer soln.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX22A500 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberY18A906 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberF21X711 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberF21X710 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberY18A907 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bicarbonate, 1M buffer soln., pH 8.0, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.