Learn More
Buerkle™ Battery Charger for Buerkle™ AccuOne/EnergyOne
Buerkle™ Battery Charger for Buerkle™ Accu/EnergyOne is the charging accessory for the lithium-ion rechargeable battery.
Brand: Buerkle™ 5900-3015
1235.00 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Description
Battery Charger for Buerkle™ Accu/EnergyOne is the correct charger for the lithium-ion rechargeable battery of the AccuOne.
Specifications
Battery Charger | |
Accu/EnergyOne Barrel Pump | |
Charger for battery Accu/EnergyOne | |
0.8 kg |
Promotions
-
Multiple Manufacturers
Everyday Lab Essentials
From wipes to waste bins, labels to liners – save on them all.Promo Code: 24132Expires: 2025-08-24
New Offer Shop Now