missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bethanechol Chloride, 100.2%
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Specifications
Specifications
Color | White |
Assay Percent Range | 100.2% |
Quantity | 5 g |
Formula Weight | 196.7 |
Physical Form | Powder |
Chemical Name or Material | Bethanechol Chloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bethanechol Chloride, 100.2% >
Spot an opportunity for improvement?Share a Content Correction