missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Betaine Free Base MP Biomedicals
Click to view available options
Quantity:
50 g
Specifications
Specifications
CAS | 107-43-7 |
Molecular Formula | C5H11NO2 |
Quantity | 50 g |
Synonym | betaine, glycine betaine, oxyneurine, lycine, trimethylglycine, abromine, trimethylglycocoll, glycylbetaine, glycocoll betaine, acidin-pepsin |
InChI Key | KWIUHFFTVRNATP-UHFFFAOYSA-N |
SMILES | C[N+](C)(C)CC(=O)[O-] |
IUPAC Name | 2-(trimethylazaniumyl)acetate |
Molecular Weight (g/mol) | 117.148 |
PubChem CID | 247 |
ChEBI | CHEBI:17750 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'