Learn More
beta-Propiolactone, 98%, pure Product Code.: 10358153
beta-Propiolactone, 98%, pure
Product Code. 10358153 Shop All Thermo Scientific Chemicals Products
100 mL, Glass bottle, 100mL
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
5mL
Product Code. 10358153
Brand: Thermo Scientific Acros 269041000
Request Bulk or Custom Format
CAS: 57-57-8 | C3H4O2 | 72.06 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Packaging Note: Glass bottle is put into a can for shipping.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57-57-8
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
MDL Number MFCD00005169
InChI Key VEZXCJBBBCKRPI-UHFFFAOYSA-N
Synonym beta-propiolactone, propiolactone, 2-oxetanone, propanolide, betaprone, 3-propanolide, 1,3-propiolactone, 3-propiolactone, beta-propionolactone, beta-propiolakton
PubChem CID 2365
ChEBI CHEBI:49073
IUPAC Name oxetan-2-one
SMILES O=C1CCO1
Specifications
Density 1.1460g/mL
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 mL
Formula Weight 72.06
Grade Pure
Chemical Name or Material β-Propiolactone, Pure
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0438090 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code269041000,269040050,269040250,269045000
Lot NumberA0444258 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code269041000,269040050,269040250,269045000
Lot NumberA0441579 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code269041000,269040050,269040250,269045000
Lot NumberA0437091 Certificate TypeCertificate of Analysis Date31/01/2025 Product Code269041000,269040050,269040250,269045000
Lot NumberA0441579 Certificate TypeCertificate of Analysis Date01/05/2024 Product Code269041000,269040050,269040250,269045000
Product Title
beta-Propiolactone, 98%, pure >