missing translation for 'onlineSavingsMsg'
Learn More

beta-III Tubulin Rabbit anti-Human, Clone: TUBB3/7089R, Novus Biologicals™

Rabbit Monoclonal Antibody

2630.00 NOK - 4855.00 NOK

Specifications

Antigen beta-III Tubulin
Clone TUBB3/7089R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18329446
View Documents
Bio-Techne
NBP3-13836-20UG
20 μg
2630.00 NOK
20µg
Please to purchase this item. Need a web account? Register with us today!
18322752
View Documents
Bio-Techne
NBP3-13836-100UG
100 μg
4855.00 NOK
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

beta-III Tubulin Monoclonal antibody specifically detects beta-III Tubulin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

beta-III Tubulin
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Biology, Cell Cycle and Replication, Cellular Markers, Neuroscience
10 mM PBS with 0.05% BSA
beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen
A synthetic peptide (aa 437-450) of human Tubulin beta 3 protein (TUBB3); coupled to KLH. (Uniprot: Q13509 )
Primary
Store at 4°C.
TUBB3/7089R
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
Q13509
10381
IgG
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.