Learn More
Bel-Art™ SP Scienceware™ Fluorescent Ruler
Useful for all fluorescent viewing and fluorescent photographic work
Brand:  Bel-Art™ F13355-0001
264.34 NOK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Product Code. 10528731
  • 284.52 NOK / Each
Everyday Lab Essentials
Use promo code 24132 at checkout
View Available Promotions
Only null left
Estimated Shipment: 20-08-2025
Description
  • The scale will glow visibly when illuminated with ultraviolet light
  • This Metric/English scale is printed on plastic with a pink phosphorescent ink
  • The scale is 0 to 15cm with millimeter increments on one edge and 0 to 6 in. divided into 0.0625 in. units on the other edge
  • Serves as a measure of nucleic acid mobility in gels
Specifications
Specifications
Ruler, Fluorescent
Plastic
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Bel-Art™ SP Scienceware™ Fluorescent Ruler > Phosphorescent ink Metric and English scales