Learn More
SP Bel-Art™ Bel Art Products Trypsin Warming Plate
Highly accurate Warming Plate, set at an optimal 37°C (±1°C), is designed to Trypsinize Cells during cell passaging
Brand:  SP Bel-Art™ F37015-3000
Product Code. 15523034
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Avoids numerous entries into the CO2 incubator which alter its atmosphere, injuring ongoing experiments in the incubated cultures
  • Increases cell viability by keeping cells at an optimal temperature during examination of cell cultures outside of the incubator
  • Can be used to maintain the warmth of reagents inside the hood
  • Built in, 15 minute count down timer keeps track of the time that the trypsin has been on the cells
  • Temperature remains uniform across the entire anodized aluminum warming plate; measures 17.8 D x 20.3 cm W (7 x 8 in.)
  • Fits easily in cell culture hoods or on the workbench
  • UV resistant polypropylene housing will not break down under germicidal UV lights
  • Water resistant and safe for cleaning with Ethanol or IPA
Specifications
Specifications
25.4 x 24.9 x 5.7 cm
220 V
37°C
24.9 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SP Bel-Art™ Bel Art Products Trypsin Warming Plate > Voltage: 220 V