Learn More
Buerkle™ Beakers with Handle and Transparent Graduations
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
5000 mL
Description
- PP (polypropylene)
- Translucent
- Autoclavable
- Graduated for measuring
Specifications
Specifications
Color | Translucent |
Diameter (Metric) Base | 50 mm |
Diameter (Metric)Top | 70 mm |
Capacity (Metric) | 250 mL |
Material | Polypropylene |
For Use With (Application) | Suitable for Foodstuff |
Height (Metric) | 120 mm |
Includes | Handle |
Quantity | 1 |
Type | Beaker |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Buerkle™ Beakers with Handle and Transparent Graduations >