Learn More
BD™ Falcon™ Roller Bottles, Easy On/Off Cap
Compatible with manual and automated filling systems
Brand:  BD Biosciences 353008
Product Code. 11537411
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Ideal for use in research and manufacturing applications involving the scale-up of mammalian cells for purposes of virus propagation and bioproduct production.
  • One-piece BD Tufrol™ polystyrene manufactured using a proprietary process that provides high impact resistance
  • Sterilized by gamma irradiation, nonpyrogenic
Custom packaging and lot control available. Contact your Fisher Scientific Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Smooth
Polystyrene
850 cm2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BD™ Falcon™ Roller Bottles, Easy On/Off Cap > Smooth Surface; 850cm2 Growth area; Easy On/Off Cap; 20/cs